Learn More
Cayman Chemical ANTIBODY DMPAC-Chol 5mg
SDP
Supplier:  Cayman Chemical 265835 MG
Encompass_Preferred
A cationic cholesterol derivative; has been used in liposome formation for gene transfection; a component of liposomes that bind to DNA in a band shift assay and protect against serum nuclease degradation of DNA at lipsome:DNA ratios from 2.5:1-10:1 w/w; reduces viability of HepG2 cells by 63% when used in liposomes at 37.5 UG/ml
Catalog No. 50-293-3896