Learn More
Cayman Chemical ANTIBODY DPQ 5mg
SDP
Supplier:  Cayman Chemical 144505 MG
Encompass_Preferred
A potent inhibitor of PARPs, inhibiting PARP1 with an IC50 value of 40 nM; about 10-fold less potent against PARP2
Catalog No. 50-296-9762