Learn More
Cayman Chemical ANTIBODY DR2313 10mg
SDP
Supplier:  Cayman Chemical 2118110 MG
Encompass_Preferred
A PARP inhibitor (IC50 = 0.2 and 0.24 µM for PARP1 and PARP2, respectively, in nuclear rat brain extracts); selective for PARP over GAPDH, ADH, or LDH; has no effect on lipid peroxidation; prevents cell death in vitro and in vivo
Catalog No. 50-298-1644