Learn More
Cayman Chemical ANTIBODY DTPA 25g
SDP
Supplier:  Cayman Chemical 3330725 G
Encompass_Preferred
A cell-impermeable metal chelating agent; chelates copper, iron, zinc, and manganese, as well as calcium, magnesium, and plutonium; has been used as a metal chelating agent in the study of ferroptosis; reduces the skeletal plutonium burden and incidence of bone tumors, as well as increases survival in a mouse model of intravenous plutonium exposure at 500 mg/kg
Catalog No. 50-294-9635