Learn More
Cayman Chemical ANTIBODY DUPA 5mg
SDP
Supplier:  Cayman Chemical 195015 MG
Encompass_Preferred
An inhibitor of PSMA (IC50 = 47 nM for the glutamate carboxypeptidase activity); has been used as a ligand to target radiolabels, cytotoxic agents, and antibody-drug conjugates to cancer cells
Catalog No. 50-298-1090