Learn More
Cayman Chemical ANTIBODY Dynasore 10mg
SDP
Supplier:  Cayman Chemical 1406210 MG
Encompass_Preferred
A cell-permeable, reversible inhibitor of dynamin 1, dynamin 2, and mitochondrial dynamin, Drp1 GTPase activity (IC50 = 15 μM); inhibits clathrin-dependent vesicle formation and endocytosis; can also augment the release of neurotransmitters and secreted cytokines
Catalog No. 50-295-3822