Learn More
Cayman Chemical ANTIBODY DyphylIn 250mg
SDP
Supplier:  Cayman Chemical 17958250 MG
Encompass_Preferred
A xanthine derivative that acts as an adenosine receptor antagonist and phosphodiesterase inhibitor and exhibits bronchodilator and vasodilator effects; also inhibits mPGES-1 (IC50 = 200 µM) and downregulates ABCG2 drug transporter expression in multidrug resistant tumor cells
Catalog No. 50-297-9718