Learn More
Cayman Chemical ANTIBODY Eg5-I 10mg
SDP
Supplier:  Cayman Chemical 1666810 MG
Encompass_Preferred
A potent inhibitor of Eg5, both in vitro and in cells (IC50s = 127 and 190 nM, respectively); blocks bipolar spindle formation and inhibits the growth of cancer cells, with an average GI50 value of 360 nM against a broad panel of tumor cells
Catalog No. 50-298-0279