Learn More
Cayman Chemical ANTIBODY ElacrIdar 50mg
SDP
Supplier:  Cayman Chemical 1812850 MG
Encompass_Preferred
A dual inhibitor of MRP-1 and BCRP; maximally inhibits MRP-1 in isolated human peripheral blood leukocytes at 200 nM; effective against both human BCRP and the mouse homolog, Bcrp1
Catalog No. 50-298-0516