Learn More
Cayman Chemical ANTIBODY ElIprodIl 50mg
SDP
Supplier:  Cayman Chemical 1839850 MG
Encompass_Preferred
A non-competitive NMDA receptor antagonist that inhibits neuronal calcium channel currents; selective for NR2B subunit-containing receptors (IC50 = 1 µM), displaying greater than 100-fold selectivity for NR2B over NR2A and NR2C subunits
Catalog No. 50-296-6525