Learn More
Cayman Chemical ANTIBODY EMA401 1mg
SDP
Supplier:  Cayman Chemical 229881 MG
Encompass_Preferred
A selective AT2 receptor antagonist (IC50s = 39.5 and 408,000 nM for rat recombinant AT2 and AT1, respectively); inhibits capsaicin-induced calcium influx in cultured hDRG neurons (IC50 = 10 nM); reduces neurite density and length in rat DRG neuronal cultures; has analgesic effects in a rat model of neuropathic pain induced by chronic constriction injury
Catalog No. 50-296-4516