Learn More
Cayman Chemical ANTIBODY EMD 638683 5mg
SDP
Supplier:  Cayman Chemical 311175 MG
Encompass_Preferred
An inhibitor of SGK1; inhibits SGK1 by 85% at 1 µM; >27-fold selective for SGK1 over a panel of 11 kinases, but does inhibit SGK2, SGK3, PRK2, and MSK1 by >50% at 1 µM; inhibits phosphorylation of the SGK1 target NDRG1 in HeLa cells (IC50 = 3.35 µM); increases radiation-induced apoptosis of Caco-2 colon carcinoma cells at 50 µM; dietary administration reduces the number of tumors in a mouse model of chemical carcinogenesis; prevents fructose and saline consumption-induced increases in systolic blood pressure in mice; decreases body weight, fasting blood glucose and HbA1c levels, and food intake in db/db diabetic mice; reduces angiotensin II-induced collagen deposition and cardiac fibrosis in mice
Catalog No. 50-298-9493