Learn More
Cayman Chemical ANTIBODY EML 425 5mg
SDP
Supplier:  Cayman Chemical 306855 MG
Encompass_Preferred
An inhibitor of p300 and CBP (IC50s = 2.9 and 1.1 µM, respectively); prevents glucose-induced cataract formation in isolated rat lens at 200 µM
Catalog No. 50-298-8718