Learn More
Cayman Chemical ANTIBODY Endothal 50mg
SDP
Supplier:  Cayman Chemical 1740450 MG
Encompass_Preferred
An aquatic plant growth regulator; reduces curly leaf pondweed appearance frequency and dry biomass in lakes at 1 mg/L annually; reduces leaf chlorophyll content in A. thaliana plants and root length in A. thaliana seedlings (IC50s = 1.36 mM and 13.9 µM, respectively) and decreases duckweed growth rate (IC50 = 10 µM); induces production of ethylene in bean leaves at 1 µM; a PP2A inhibitor (IC50 = 19-50 nM); toxic to mice (LD50 = 5-50 mg/kg)
Catalog No. 50-296-5459