Learn More
Cayman Chemical ANTIBODY ENMD-2076 1mg
SDP
Supplier:  Cayman Chemical 229561 MG
Encompass_Preferred
A multi-kinase inhibitor; inhibits FLT3, RET, and Aurora A kinase (IC50s = 1.86, 10.4, and 14 nM, respectively); inhibits additional kinases involved in angiogenesis (IC50s = 50s = via oral gavage); decreases tumor vascular permeability and perfusion and inhibits tumor growth in human TNBC and colorectal cancer mouse xenograft models of (100 mg/kg via oral gavage)
Catalog No. 50-296-4410