Learn More
Cayman Chemical ANTIBODY EPZ020411 5mg
SDP
Supplier:  Cayman Chemical 191605 MG
Encompass_Preferred
An inhibitor of PRMT6 (IC50 = 10 nM) that less potently targets PRMT1 and PRMT8 (IC50s = 119 and 223 nM, respectively); dose-dependently decreases methylation of the PRMT6 substrate H3R2 in A375 cells transiently expressing PRMT6 (IC50 = 0.637 µM)
Catalog No. 50-298-3067