Learn More
Cayman Chemical ANTIBODY EquIsetIn 1mg
SDP
Supplier:  Cayman Chemical 181961 MG
Encompass_Preferred
An HIV-1 integrase inhibitor; inhibits Dnp-stimulated ATPase activity of rat liver mitochondria and mitoplasts (IC50 = ~8 nM per mg of protein for both); inhibits ADP-stimulated respiration and the mitochondrial transport of ATP, inorganic phosphate, and succinate; phytotoxic
Catalog No. 50-296-7985