Learn More
Cayman Chemical ANTIBODY ErastIn 5mg
SDP
Supplier:  Cayman Chemical 177545 MG
Encompass_Preferred
A ferroptosis inducer; induces ferroptotic cell death in vitro, an effect that can be blocked by ferroptosis inhibitors; inhibits cystine uptake by the system xc- cystine-glutamate transporter in HT-1080 fibrosarcoma and Calu-1 lung carcinoma cells at 5 µM; inhibits glutamate release via the cystine-glutamate transporter in an enzyme-coupled fluorescence assay (EC50 = 1.2 µM); selectively induces cell death in cells expressing the SV40 small T oncoprotein and oncogenic Ras (IC50s = 1.25-5 UG/ml)
Catalog No. 50-297-9878