Learn More
Cayman Chemical ANTIBODY ErIbulIn 10mg
SDP
Supplier:  Cayman Chemical 1735110 MG
Encompass_Preferred
An inhibitor of microtubule assembly; inhibits the proliferation of U937 human lymphoma cells, as well as induces cell cycle arrest at the G2/M phase and, subsequently, apoptosis in the same cells at 100 nM; reduces tumor growth in a PDX mouse model of cutaneous squamous cell carcinoma at 1.5 mg/kg per week; reduces tumor growth and increases survival in an intracerebral U87MG mouse xenograft model of glioblastoma at 0.5 mg/kg three times per week
Catalog No. 50-296-7651