Learn More
Cayman Chemical ANTIBODY ETP-46464 10mg
SDP
Supplier:  Cayman Chemical 1980910 MG
Encompass_Preferred
An ATR inhibitor (IC50 = 25 nM) that also inhibits mTOR and DNA-PK (IC50s = 0.6 and 36 nM, respectively); generates replicative stress, leading to chromosomal breakage in the presence of conditions that stall replication forks
Catalog No. 50-298-4943