Learn More
Cayman Chemical ANTIBODY Eucalyptol 10mg
SDP
Supplier:  Cayman Chemical 2218310 MG
Encompass_Preferred
A bicyclic monoterpene; inhibits ionomycin- and PMA-induced TNF-α, IL-1β, IL-4, and IL-5 production by primary human lymphocytes at 10 μM; decreases LPS-induced mucus production by primary human nasal turbinate slices at 10 μM; decreases carrageenan-induced hind paw edema in rats and reduces the time spent licking the hind paw in a formalin-induced nociception test in mice at 400 mg/kg; inhibits A. aegypti mosquitoes from feeding on anesthetized gerbils when applied topically at 10% and from laying eggs in an ovipositional bioassay at 1% in standing water
Catalog No. 50-298-7291