Learn More
Cayman Chemical ANTIBODY Exemestn 25mg
SDP
Supplier:  Cayman Chemical 1500825 MG
Encompass_Preferred
An irreversible inhibitor of aromatase (Ki = 26 nM); is selective for aromatase over 5α-reductase at 30 µM; decreases tumor proliferation in rats with DMBA-induced mammary tumors at 3-100 mg/kg per day, s.c.
Catalog No. 50-296-8731