Learn More
Cayman Chemical ANTIBODY FAUC-365 1mg
SDP
Supplier:  Cayman Chemical 204571 MG
Encompass_Preferred
A dopamine D3 receptor antagonist (Ki = 0.5 nM); selective for dopamine D3 receptors over dopamine D1, D2L, D2S, and D4 receptors (Kis = 8.8, 3.6, 2.6, and 0.34 µM, respectively) as well as 5-HT1A and 5-HT2 receptors (Kis = 0.36 and 2 µM, respectively); prevents memory impairment in the novel object recognition test in DAT-KD mice when administered prior to object learning from 1-10 mg/kg
Catalog No. 50-298-1126