Learn More
Cayman Chemical ANTIBODY FDW028 25mg
SDP
Supplier:  Cayman Chemical 3954325 MG
Encompass_Preferred
A FUC8 inhibitor; binds to FUC8 (Kd = 5.486 µM) and inhibits core fucosylation in HCT-8 and SW480 CRC cells; induces degradation of B7-H3 via CMA in the same cells at 50 µM; reduces tumor volume and increases survival in an SW480 mouse xenograft model at 10 and 20 mg/kg
Catalog No. 50-291-9183