Learn More
Cayman Chemical ANTIBODY FGFR4-IN-1 10mg
SDP
Supplier:  Cayman Chemical 2134210 MG
Encompass_Preferred
An inhibitor of FGFR4 (IC50 = 1.3 nM); selective for FGFR4 over FGFR1 and a panel of 36 kinases (IC50s = >10 µM for all); inhibits proliferation of Hep3B and Huh7 hepatic cancer cells with IC50 values of 1.1 and 2.5 nM, respectively; reduces tumor growth by 62.7 and 70.8% in a Huh7 mouse xenograft model at 30 and 100 mg/kg per day, respectively
Catalog No. 50-298-5034