Learn More
Cayman Chemical ANTIBODY FH535 5mg
SDP
Supplier:  Cayman Chemical 180015 MG
Encompass_Preferred
An inhibitor of β-catenin/Tcf-mediated transcription and an antagonist of ligand-dependent activation of PPARγ and PPARδ; blocks the recruitment of β-catenin and GRIP1 to PPARγ and PPARδ; selectively toxic to some carcinoma cell lines expressing the Wnt/β-catenin pathway
Catalog No. 50-297-9725