Learn More
Cayman Chemical ANTIBODY FIN56 25mg
SDP
Supplier:  Cayman Chemical 2518025 MG
Encompass_Preferred
An inducer of ferroptosis; induces ferroptosis in BJ cells overexpressing oncogenic HRASG12V via acetyl-CoA carboxylase-dependent reduction in expression of GPX4 protein; binds to and activates squalene synthase in a GPX4 degradation-independent manner
Catalog No. 50-293-3933