Learn More
Cayman Chemical ANTIBODY FIpronIl 50mg
SDP
Supplier:  Cayman Chemical 2412850 MG
Encompass_Preferred
An insecticide; a selective antagonist of insect GABA receptors (IC50s = 28 and 1,660 nM for cockroach and rat receptors, respectively); inhibits desensitizing and non-desensitizing glutamate-induced chloride currents in cockroach neurons (IC50s = 800 and 10 nM, respectively); induces CYP1A1/2, CYP2B1/2, and CYP3A1/2 activity in isolated rat liver microsomes
Catalog No. 50-293-3294