Learn More
Cayman Chemical ANTIBODY FlumazenIl 100mg
SDP
Supplier:  Cayman Chemical 14252100 MG
Encompass_Preferred
A GABAA receptor antagonist (IC50 = 2 nM in a radioligand binding assay using rat cortical synaptosomes); decreases the amplitude of electrically stimulated population spikes in rat hippocampal CA1 pyramidal neurons; increases the number of entries into the open arms of the elevated plus maze in BALB/c, but not C57BL/6, mice from 0.1-1,000 UG/kg; prevents allopregnanolone-induced reduction of burying behavior in ovariectomized rats at 5 and 10 mg/kg
Catalog No. 50-295-4484