Learn More
Cayman Chemical ANTIBODY FlumequIn 1g
SDP
Supplier:  Cayman Chemical 216451 G
Encompass_Preferred
A fluoroquinolone antibiotic; active against S. aureus, S. pyogenes, B. subtilis, E. coli, P. aeruginosa, S. faecalis, and K. pneumoniae (MICs = 1-100 UG/ml); active against field isolates of B. hyodysenteriae (MICs = 6.25-200 UG/ml); increases survival in rat models of P. vulgaris-induced urinary tract infection and P. mirabilis-induced prostatitis at 50 mg/kg
Catalog No. 50-298-6122