Learn More
Cayman Chemical ANTIBODY Fluo-3 AM 1mg
SDP
Supplier:  Cayman Chemical 149601 MG
Encompass_Preferred
A cell-permeable AM ester of the fluorescent calcium indicator fluo-3
Catalog No. 50-297-0812