Learn More
Cayman Chemical ANTIBODY FngolImod 50mg
SDP
Supplier:  Cayman Chemical 1197550 MG
Encompass_Preferred
An immune modulator and a prodrug form of FTY720 phosphate; decreases the number of circulating lymphocytes in mice at 1 mg/kg; increases skin allograft survival in rats from 0.3-3 mg/kg; prevents disease development in a rat model of EAE; inhibits S1P lyase from 0.3-30 µM
Catalog No. 50-295-4921