Learn More
Cayman Chemical ANTIBODY FurosemIde 1g
SDP
Supplier:  Cayman Chemical 172731 G
Encompass_Preferred
A loop diuretic and an inhibitor of NKCC1 and NKCC2 (Kis = ~10 µM for both); increases diuresis in beagle dogs at 0.1 mg/kg; reduces ventricular collagen deposition and fibrosis in a rat model of dilated cardiomyopathy at 30 mg/kg
Catalog No. 50-297-9433