Learn More
Cayman Chemical ANTIBODY G10 5mg
SDP
Supplier:  Cayman Chemical 223535 MG
Encompass_Preferred
An indirect activator of STING signaling; induces IRF3- and IFN-dependent transcription in human fibroblasts, PBMCs, and HUMECs; prevents replication of chikungunya and Venezuelan equine encephalitis alphaviruses (IC90s = 8.01 and 24.57 µM, respectively); increases phosphorylation of IRF3 when used at a concentration of 100 µM but not in cells lacking STING
Catalog No. 50-298-7712