Learn More
Cayman Chemical ANTIBODY GalactItol 250g
SDP
Supplier:  Cayman Chemical 25023250 G
Encompass_Preferred
A reduced form of galactose; a toxic metabolite of galactose in humans; accumulates in the lens of patients with galactokinase deficiency-induced galactosemia, leading to cataract formation; accumulates in a transgenic mouse model of galactokinase deficiency but does not lead to cataract formation
Catalog No. 50-293-3957