Learn More
Cayman Chemical ANTIBODY GDC-0152 1mg
SDP
Supplier:  Cayman Chemical 178101 MG
Encompass_Preferred
A potent inhibitor of specific IAPs that binds baculovirus IAP repeat (BIR) domains (Kis = 28, 14, 17, and 43 nM for XIAP BIR3, NL-IAP BIR, BIRC2 (c-IAP1) BIR3, and BIRC3 (c-IAP2) BIR3, respectively); induces activation of caspases, decreasing the viability of cancer cells without affecting normal cells
Catalog No. 50-296-6665