Learn More
Cayman Chemical ANTIBODY GDC-0623 25mg
SDP
Supplier:  Cayman Chemical 1947925 MG
Encompass_Preferred
A potent, ATP-uncompetitive inhibitor of MEK1 (Ki = 0.13 nM in the presence of ATP); inhibits the proliferation of A375 BRAF(V600E) and HCT116 KRAS(G13D)-mutant cancer cell lines (EC50 = 7 and 42 nM, respectively); upregulates pro-apoptotic BIM
Catalog No. 50-298-3228