Learn More
Cayman Chemical ANTIBODY GDC-6036 500ug
SDP
Supplier:  Cayman Chemical 41852500 UG
Encompass_Preferred
A covalent inhibitor of K-RasG12C; decreases tumor volume in a PaCa-2 pancreatic cancer mouse xenograft model at 1 to 100 mg/kg per day
Catalog No. 50-294-3822