Learn More
Cayman Chemical ANTIBODY Gestoden 250mg
SDP
Supplier:  Cayman Chemical 18217250 MG
Encompass_Preferred
A progestin; selectively binds to the progesterone receptor over the estrogen receptor; induces transcriptional activity mediated by the progesterone receptor, and, weakly, the androgen receptor in reporter assays; increases the growth of MCF-7 cells at 0.1-1 µM; reduces tumor growth in an estrogen-dependent model of DMBA-induced rat mammary tumors at 0.1 mg/kg
Catalog No. 50-296-6725