Learn More
Cayman Chemical ANTIBODY GGTI 2418 5mg
SDP
Supplier:  Cayman Chemical 287145 MG
Encompass_Preferred
An inhibitor of GGTase I (IC50 = 9.5 nM); selective for GGTase I over FTase (IC50 = 53,000 nM); reduces tumor growth in an MDA-MB-231 breast cancer mouse xenograft model at 100 mg/kg
Catalog No. 50-293-7073