Learn More
Cayman Chemical ANTIBODY GIT 27 25mg
SDP
Supplier:  Cayman Chemical 1629325 MG
Encompass_Preferred
An immunomodulatory compound that inhibits inflammatory antigen presentation both in vitro and in preclinical in vivo models of autoimmune diseases; targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, macrophage migration inhibitory factor, and inducible nitric oxide synthase-mediated nitric oxide generation
Catalog No. 50-297-2545