Learn More
Cayman Chemical ANTIBODY GKT137831 5mg
SDP
Supplier:  Cayman Chemical 177645 MG
Encompass_Preferred
A dual inhibitor of both NOX1 and NOX4 with Ki values in the range of 100 to 150 nM in cell-free assays of ROS production; displays good oral bioavailability with high plasma concentrations in vivo and attenuates liver fibrosis in mice
Catalog No. 50-296-6719