Learn More
Cayman Chemical ANTIBODY GlycIteIn 25mg
SDP
Supplier:  Cayman Chemical 1416225 MG
Encompass_Preferred
An O-methylated isoflavone with weak estrogenic activity (IC50 = 3.94 μM) that has been used in research related phytoestrogen therapeutic activity applied to bone formation, cardiovascular disease, estrogen-dependent cancer, and Alzheimer’s disease
Catalog No. 50-295-2924