Learn More
Cayman Chemical ANTIBODY Go 6976 500ug
SDP
Supplier:  Cayman Chemical 13310500 UG
Encompass_Preferred
A PKCα and PKCβ1 inhibitor with IC50 values of 2.3 and 6.2 nM, respectively; also inhibits the checkpoint kinases Chk1/2 and abrogates DNA damage-induced cell cycle arrest, leading to cytotoxicity in p53 mutant tumor cells
Catalog No. 50-295-2318