Learn More
Cayman Chemical ANTIBODY GomIsIn C 5mg
SDP
Supplier:  Cayman Chemical 186435 MG
Encompass_Preferred
Scavenges oxygen radicals and reduces the production of ROS by inhibiting neutrophil activity; attenuates respiratory burst in rat peripheral neutrophils via suppression of NADPH oxidase (39% inhibition at 30 UG/ml) and inhibition of cytosolic calcium release from intracellular stores (36% inhibition at 50 UG/ml)
Catalog No. 50-296-4999