Learn More
Cayman Chemical ANTIBODY GS-7340 50mg
SDP
Supplier:  Cayman Chemical 2621450 MG
Encompass_Preferred
A prodrug from of tenofovir; reduces HIV-1 viral replication in MT-2 cells (EC50 = 5 nM)
Catalog No. 50-293-4067