Learn More
Cayman Chemical ANTIBODY GSA 10 5mg
SDP
Supplier:  Cayman Chemical 161645 MG
Encompass_Preferred
A SMO agonist that can promote the differentiation of multipotent mesenchymal progenitor cells into osteoblasts (EC50 = 1.2 µM); used to characterize a novel SMO active site since it does not recognize the classic cyclopamine binding site utilized by other SMO agonists
Catalog No. 50-297-0713