Learn More
Cayman Chemical ANTIBODY GSK137647A 5mg
SDP
Supplier:  Cayman Chemical 177125 MG
Encompass_Preferred
A selective agonist of GPR120 (EC50s = 0.5, 0.63, and 0.79 µM for human, mouse, and rat receptors, respectively); dose-dependently stimulates insulin secretion by mouse insulinoma MIN6 cells under high glucose conditions
Catalog No. 50-297-9560