Learn More
Cayman Chemical ANTIBODY GSK180736A 25mg
SDP
Supplier:  Cayman Chemical 2618225 MG
Encompass_Preferred
A ROCK1 and GRK2 inhibitor (IC50s = 14 and 0.77 µM, respectively); selective for GRK2 over GRK1, GRK5, and PKA (IC50 = >100 µM for all); selective for ROCK1 over RSK1 and p20S6K (IC50s = 3,100 and 2,850 nM, respectively); increases maximum contractility in isolated mouse cardiomyocytes at 1 µM
Catalog No. 50-293-5198