Learn More
Cayman Chemical ANTIBODY GSK2636771 25mg
SDP
Supplier:  Cayman Chemical 1738025 MG
Encompass_Preferred
An orally bioavailable inhibitor of PI3K p110β that decreases cell viability and Akt phosphorylation in p100β-dependent PTEN-deficient PC-3 prostate and BT549 and HCC70 breast cancer cell lines
Catalog No. 50-296-8326